Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138352.1 | NP_612361.1 | 1448 | |||
GenBank mRNA | AY453840.1 | - | 1448 | |||
BC007384.2 | - | 4, 5 | 542 | |||
BC030129.2 | - | 4, 5 | 721 | |||
BC065477.1 | - | 4, 5 | 689 | |||
BC080588.1 | - | 4, 5 | 542 |