Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC90768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027107.2 | 4 | 1405 | |||
GenBank mRNA | AK024300.1 | - | 4 | 1356 | ||
BC033172.1 | - | 1 | 857 |