Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033549.4 | NP_291027.3 | 6 | 3220 | ||
NM_201627.2 | NP_963921.1 | 8 | 2280 | |||
NR_045218.1 | 7 | 2257 | ||||
XM_006714929.2 | XP_006714992.1 | 7 | 2642 | |||
XM_006714930.2 | XP_006714993.1 | 7 | 2636 | |||
XM_006714931.3 | XP_006714994.1 | 8 | 2274 | |||
XM_006714932.3 | XP_006714995.1 | 8 | 2284 | |||
GenBank mRNA | AB063180.1 | - | 8 | 1306 | ||
AB100366.1 | - | 6 | 2482 | |||
AB100367.1 | - | 8 | 1543 | |||
AF258579.1 | - | 2 | 1576 | |||
AK027601.1 | - | 8 | 2253 | |||
AK074052.1 | - | 2 | 5271 | |||
AL137593.1 | - | 4 | 1931 | |||
AL832960.1 | - | 5 | 2297 | |||
AX775789.1 | - | 6 | 1959 | |||
AX775791.1 | - | 6 | 1959 | |||
AX775793.1 | - | 8 | 2253 | |||
AX775797.1 | - | 6 | 2251 | |||
AX775799.1 | - | 6 | 3193 | |||
AX775801.1 | - | 5 | 3393 | |||
AX775803.1 | - | 5 | 2451 | |||
BC004956.1 | - | 7 | 1462 | |||
BC009762.2 | - | 2 | 1254 | |||
BC018765.1 | - | 6 | 2251 | |||
BC071887.1 | - | 7 | 1462 | |||
BC090953.1 | - | 6 | 2761 | 6017628 | ||
BC111568.1 | - | 3 | 307 | |||
CA430637.1 | - | 1 | 458 |