Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002862.2 | NP_001002862.1 | 3 | 232 | ||
NM_001135751.1 | NP_001129223.1 | 3 | 232 | |||
NM_198440.3 | NP_940842.2 | 3 | 232 | |||
XM_011530505.2 | XP_011528807.1 | 3 | 342 | |||
XM_011530506.2 | XP_011528808.1 | 3 | 340 | |||
XM_017029078.1 | XP_016884567.1 | 3 | 388 | |||
XM_017029079.1 | XP_016884568.1 | 3 | 546 | |||
XM_017029080.1 | XP_016884569.1 | 3 | 504 | |||
XM_017029081.1 | XP_016884570.1 | 3 | 361 | |||
XM_017029082.1 | XP_016884571.1 | 2 | 237 | |||
GenBank mRNA | AK096976.1 | - | 2 | 237 | ||
AK125830.1 | - | 2 | 342 | |||
AL389876.1 | - | 3 | 232 | |||
BC057830.1 | - | 3 | 225 | 30338943 | ||
BM792257.1 | - | 1 | 39 | |||
CR456372.1 | - | 3 | 226 |