Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002862.2 | NP_001002862.1 | 4 | 319 | ||
NM_001135751.1 | NP_001129223.1 | 4 | 319 | |||
NM_198440.3 | NP_940842.2 | 4 | 319 | |||
XM_011530505.2 | XP_011528807.1 | 4 | 429 | |||
XM_011530506.2 | XP_011528808.1 | 4 | 427 | |||
XM_017029078.1 | XP_016884567.1 | 4 | 475 | |||
XM_017029079.1 | XP_016884568.1 | 4 | 633 | |||
XM_017029080.1 | XP_016884569.1 | 4 | 591 | |||
XM_017029081.1 | XP_016884570.1 | 4 | 448 | |||
XM_017029082.1 | XP_016884571.1 | 3 | 324 | |||
GenBank mRNA | AK096976.1 | - | 3 | 324 | ||
AK125830.1 | - | 3 | 429 | |||
AL389876.1 | - | 4 | 319 | |||
AW135031.1 | - | 1 | 22 | |||
BC057830.1 | - | 4 | 312 | 30338943 | ||
BM792257.1 | - | 2 | 126 | |||
CR456372.1 | - | 4 | 313 |