Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012967.2 | NP_001012985.2 | 13 | 1866 | ||
NM_001291510.1 | NP_001278439.1 | 13 | 1866 | |||
XM_005263341.4 | XP_005263398.2 | 13 | 1849 | |||
XM_005263342.2 | XP_005263399.2 | 13 | 1866 | |||
XM_005263343.2 | XP_005263400.2 | 13 | 1832 | |||
XM_005263345.4 | XP_005263402.2 | 13 | 1845 | |||
XM_006714408.2 | XP_006714471.1 | 14 | 1916 | |||
XM_011532402.2 | XP_011530704.1 | 14 | 2139 | |||
XM_011532403.2 | XP_011530705.1 | 14 | 1974 | |||
XM_011532404.1 | XP_011530706.1 | 13 | 1866 | |||
XM_011532405.1 | XP_011530707.1 | 13 | 1866 | |||
XM_011532406.1 | XP_011530708.1 | 13 | 1866 | |||
XM_011532409.2 | XP_011530711.1 | 1 | 20 | |||
XM_017008829.1 | XP_016864318.1 | 13 | 1832 | |||
XM_017008830.1 | XP_016864319.1 | 13 | 1866 | |||
XM_017008831.1 | XP_016864320.1 | 1 | 29 | |||
GenBank mRNA | AK055595.1 | - | 4 | 365 | ||
AK092461.1 | - | 7 | 802 | |||
CR933601.1 | - | 13 | 1840 | |||
KR632542.1 | - | 11 | 1618 |