Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 6, 7 | 1136 | ||
NM_033544.2 | NP_291022.2 | 7, 8 | 1220 | |||
GenBank mRNA | BC064606.1 | - | 4, 5 | 1147 | ||
BC113826.1 | - | 4, 5 | 544 | |||
BC140708.1 | - | 6, 7 | 1134 | 9021576 | ||
BC140709.1 | - | 6, 7 | 1134 | 9021577 |