Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 5 | 398 | ||
NM_145345.2 | NP_663320.2 | 9 | 878 | |||
NM_183008.2 | NP_892120.2 | 10 | 883 | |||
GenBank mRNA | AF520989.1 | - | 8 | 587 | ||
AF521017.1 | - | 8 | 588 | |||
AF521018.1 | - | 8 | 575 | |||
AK074358.1 | - | 5 | 375 | |||
AK090792.1 | - | 5 | 376 | |||
AK093258.1 | - | 10 | 827 | |||
AK094198.1 | - | 5 | 330 | |||
AK097368.1 | - | 9 | 976 | |||
AK291503.1 | - | 8 | 583 | |||
AK293910.1 | - | 3 | 222 | |||
AK296179.1 | - | 7 | 485 | |||
AY007163.1 | - | 6 | 342 | |||
BC038106.1 | - | 9 | 695 | 5269682 | ||
BX647774.1 | - | 9 | 691 | |||
BX648412.1 | - | 8 | 592 | |||
CR749413.1 | - | 9 | 689 |