Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 6 | 695 | |||
GenBank mRNA | AK098201.1 | - | 6 | 722 | ||
AL157851.1 | - | 6 | 680 | |||
BC007736.2 | - | 6 | 695 | |||
BC014647.1 | - | 6 | 693 | |||
BC143340.1 | - | 6 | 783 |