Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 3 | 414 | ||
NM_178312.2 | NP_842564.1 | 3 | 342 | |||
XM_005260863.4 | XP_005260920.1 | 3 | 769 | |||
XM_005260864.4 | XP_005260921.1 | 4 | 922 | |||
XM_005260865.4 | XP_005260922.1 | 4 | 922 | |||
XM_011529391.2 | XP_011527693.1 | 3 | 336 | |||
XM_011529392.2 | XP_011527694.1 | 4 | 861 | |||
XM_011529393.2 | XP_011527695.1 | 3 | 708 | |||
XM_017028126.1 | XP_016883615.1 | 3 | 331 | |||
GenBank mRNA | BC040904.1 | - | 3 | 414 | 5744021 | |
L20491.1 | - | 3 | 342 | |||
L20492.1 | - | 2 | 392 |