Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017971.2 | NP_001017971.1 | 10 | 1851 | ||
XM_011543719.2 | XP_011542021.1 | 9 | 1938 | |||
XM_011543720.2 | XP_011542022.1 | 8 | 1861 | |||
XM_011543723.2 | XP_011542025.1 | 10 | 2498 | |||
XM_017010031.1 | XP_016865520.1 | 10 | 2029 | |||
XM_017010032.1 | XP_016865521.1 | 11 | 2199 | |||
XM_017010033.1 | XP_016865522.1 | 10 | 1662 | |||
XM_017010034.1 | XP_016865523.1 | 9 | 1987 | |||
XM_017010036.1 | XP_016865525.1 | 9 | 1962 | |||
XM_017010037.1 | XP_016865526.1 | 8 | 1899 | |||
XM_017010041.1 | XP_016865530.1 | 11 | 2576 | |||
XM_017010042.1 | XP_016865531.1 | 12 | 2482 | |||
XM_017010043.1 | XP_016865532.1 | 11 | 2565 | |||
XM_017010044.1 | XP_016865533.1 | 10 | 2524 | |||
XM_017010045.1 | XP_016865534.1 | 10 | 2076 | |||
XM_017010046.1 | XP_016865535.1 | 10 | 2157 | |||
XM_017010047.1 | XP_016865536.1 | 9 | 2446 | |||
GenBank mRNA | AK022625.1 | - | 10 | 1851 | ||
BC093982.1 | - | 4 | 581 | 7939827 | ||
BC112191.1 | - | 4 | 581 | 8327659 |