Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 1176 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 960 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 1079 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 978 | |||
NM_178014.3 | NP_821133.1 | 4 | 1092 | |||
GenBank mRNA | AB062393.1 | - | 4 | 918 | ||
AF070561.1 | - | 4 | 913 | |||
AF070593.1 | - | 4 | 913 | |||
AF070600.1 | - | 4 | 930 | |||
AF141349.1 | - | 4 | 930 | |||
AK297638.1 | - | 4 | 560 | |||
AK298889.1 | - | 4 | 982 | |||
AK302195.1 | - | 3 | 422 | |||
AK302314.1 | - | 4 | 1045 | |||
AK303227.1 | - | 3 | 679 | |||
AK309684.1 | - | 2 | 534 | |||
AK316265.1 | - | 4 | 849 | |||
BC001896.1 | - | 1 | 474 | |||
BC001938.1 | - | 4 | 917 | 2820585 | ||
BC002347.2 | - | 4 | 922 | 2964461 | ||
BC005838.2 | - | 4 | 917 | 2820585 | ||
BC007605.1 | - | 4 | 922 | 3350604 | ||
BC013374.2 | - | 4 | 913 | 3946253 | ||
BC015889.1 | - | 1 | 254 | |||
BC019924.1 | - | 4 | 910 | 4998571 | ||
BC020946.1 | - | 4 | 951 | 4795692 | ||
BC021909.1 | - | 4 | 912 | 3509417 | ||
BC029323.1 | - | 1 | 275 | |||
BC070326.1 | - | 4 | 928 | 6698679 | ||
BC103746.1 | - | 4 | 935 | 6503977 | ||
DQ457058.1 | - | 1 | 348 | |||
V00599.1 | - | 3 | 765 |