Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJAP1 SIRNA Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
PILT, TJP4
Chromosome Location:
Chr.6: 43477523 - 43506557 on Build GRCh38
UniGene ID:
Hs.520145
Species:
Human
siRNA ID
HSS150741
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146016.1
NP_001139488.1
11
1770
NM_001146017.1
NP_001139489.1
10
1735
NM_001146018.1
NP_001139490.1
11
1797
NM_001146019.1
NP_001139491.1
10
1740
NM_001146020.1
NP_001139492.1
9
1591
NM_080604.2
NP_542171.2
10
1792
NR_027394.1
10
1657
XM_006715249.1
XP_006715312.1
11
1748
XM_006715250.3
XP_006715313.1
12
1767
XM_006715251.2
XP_006715314.1
11
1775
XM_006715252.2
XP_006715315.1
12
1835
XM_006715254.3
XP_006715317.1
11
1673
XM_006715256.1
XP_006715319.1
11
1670
XM_006715257.2
XP_006715320.1
11
1724
XM_006715259.3
XP_006715322.1
10
1574
XM_006715260.1
XP_006715323.1
11
1691
XM_006715261.1
XP_006715324.1
10
1595
XM_006715262.2
XP_006715325.1
11
1920
XM_006715263.2
XP_006715326.1
10
1630
XM_006715265.1
XP_006715328.1
10
1640
XM_006715266.1
XP_006715329.1
9
1548
XM_006715267.2
XP_006715330.1
11
1759
XM_006715268.2
XP_006715331.1
11
1756
XM_006715269.2
XP_006715332.1
11
1811
XM_006715270.1
XP_006715333.1
10
1729
XM_011514995.1
XP_011513297.1
12
1810
XM_011514996.1
XP_011513298.1
11
1730
XM_017011488.1
XP_016866977.1
10
1648
XM_017011489.1
XP_016866978.1
11
1945
XM_017011490.1
XP_016866979.1
10
1718
XM_017011491.1
XP_016866980.1
11
1754
XM_017011492.1
XP_016866981.1
12
1883
XM_017011493.1
XP_016866982.1
12
1881
GenBank mRNA
AJ420477.1
-
2
891
AK024269.1
-
10
1742
AK074241.1
-
5
1186
AK094907.1
-
3
1598
AK123398.1
-
7
4300
AK127338.1
-
9
2853
AK225787.1
-
10
1693
AK309119.1
-
10
1610
AL117398.1
-
4
2224
BC046239.1
-
11
1690
4906152
BC064401.1
-
10
1671
5539930
CR749480.1
-
8
4121
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-nbn7j:80/100.66.77.222:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline