Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135862.2 | NP_001129334.1 | 7 | 839 | ||
NM_001242450.1 | NP_001229379.1 | 6 | 879 | |||
NM_001284296.1 | NP_001271225.1 | 7 | 925 | |||
NM_138415.4 | NP_612424.1 | 6 | 1010 | |||
NR_134535.1 | 6 | 794 | ||||
XM_006724122.1 | XP_006724185.1 | 6 | 1019 | |||
XM_011529855.1 | XP_011528157.1 | 6 | 1019 | |||
GenBank mRNA | AK091480.1 | - | 7 | 925 | ||
AK092243.1 | - | 7 | 839 | |||
AK297317.1 | - | 6 | 872 | |||
AK299836.1 | - | 6 | 835 | |||
BC012187.1 | - | 6 | 1010 | 4634631 | ||
CR456448.1 | - | 6 | 766 | |||
KU178842.1 | - | 6 | 859 | |||
KU178843.1 | - | 6 | 733 | |||
KU178844.1 | - | 6 | 733 |