Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP0P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002775.2 | 5 | 1212 | |||
GenBank mRNA | AK095332.1 | - | 5 | 1212 | ||
AY672101.1 | - | 1 | 69 |