Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 855 | ||
NM_181268.2 | NP_851785.2 | 4 | 839 | |||
NM_181269.2 | NP_851786.1 | 4 | 560 | |||
NM_181270.2 | NP_851787.1 | 4 | 544 | |||
NM_181271.2 | NP_851788.1 | 3 | 419 | |||
NM_181272.2 | NP_851789.1 | 3 | 401 | |||
NM_181283.2 | NP_851800.1 | 3 | 385 | |||
NM_181296.2 | NP_851813.1 | 2 | 286 | |||
GenBank mRNA | AF278576.1 | - | 4 | 459 | ||
AF278577.1 | - | 4 | 475 | |||
AY174118.1 | - | 4 | 564 | |||
AY174119.1 | - | 4 | 548 | |||
AY174120.1 | - | 3 | 423 | |||
AY174121.1 | - | 3 | 405 | |||
AY174122.1 | - | 3 | 389 | |||
AY174123.1 | - | 3 | 560 | |||
AY174124.1 | - | 3 | 544 | |||
AY174125.1 | - | 5 | 698 | |||
AY174126.1 | - | 5 | 682 | |||
AY174128.1 | - | 4 | 528 | |||
AY174129.1 | - | 5 | 551 | |||
AY174130.1 | - | 5 | 723 | |||
AY174131.1 | - | 3 | 380 | |||
AY174132.1 | - | 3 | 364 | |||
AY174133.1 | - | 2 | 290 | |||
AY174134.1 | - | 4 | 610 | |||
AY174135.1 | - | 5 | 788 | |||
AY174136.1 | - | 5 | 772 | |||
AY174137.1 | - | 4 | 590 | |||
AY174138.1 | - | 4 | 574 | |||
BC057852.1 | - | 4 | 855 | 5273108 | ||
DQ309766.1 | - | 5 | 673 | |||
DQ355985.1 | - | 3 | 411 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 428 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 508 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 587 |