Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135911.1 | NP_001129383.1 | 1 | 242 | ||
NM_052880.4 | NP_443112.2 | 1 | 242 | |||
GenBank mRNA | AF528079.1 | - | 1 | 187 | ||
AF528080.1 | - | 1 | 187 | |||
AK074688.1 | - | 1 | 188 | |||
AK226089.1 | - | 1 | 122 | |||
AK299397.1 | - | 1 | 196 | |||
AK300990.1 | - | 1 | 166 | |||
BC011049.2 | - | 1 | 187 | 4180134 | ||
BQ718653.1 | - | 1 | 220 | |||
CR456340.1 | - | 1 | 82 | |||
DC412134.1 | - | 1 | 242 | |||
GU135609.1 | - | 1 | 103 |