Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135911.1 | NP_001129383.1 | 5 | 711 | ||
NM_052880.4 | NP_443112.2 | 6 | 790 | |||
GenBank mRNA | AF528080.1 | - | 6 | 735 | ||
AK074688.1 | - | 6 | 736 | |||
AK093768.1 | - | 3 | 1350 | |||
AK226089.1 | - | 6 | 670 | |||
AK299397.1 | - | 5 | 665 | |||
AK300990.1 | - | 7 | 648 | |||
BC011049.2 | - | 6 | 735 | 4180134 | ||
BC041903.1 | - | 4 | 1087 | |||
BE299406.1 | - | 4 | 418 | |||
CR456340.1 | - | 6 | 630 | |||
GU135609.1 | - | 4 | 414 |