Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052888.2 | NP_443120.2 | 1 | 65 | ||
GenBank mRNA | AJ314647.1 | - | 1 | 52 | ||
AK302740.1 | - | 1 | 196 | |||
AK310287.1 | - | 1 | 93 | |||
AY386261.1 | - | 2 | 103 | |||
BC117373.1 | - | 1 | 104 | 40125924 | ||
BC143578.1 | - | 1 | 104 | |||
BC143581.1 | - | 1 | 104 | 9052092 |