Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001525.1 | 1 | 30 |
Gene Symbol: TTTY4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002178.1 | 1 | 30 |
Gene Symbol: TTTY4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002177.1 | 1 | 30 |