Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 2 | 219 | ||
NM_145326.2 | NP_663299.2 | 2 | 219 | |||
GenBank mRNA | AK092178.1 | - | 2 | 205 | ||
BC006408.1 | - | 2 | 182 | |||
BX648842.1 | - | 2 | 209 | |||
DA804007.1 | - | 2 | 219 |
Gene Symbol: ZNF430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172671.1 | NP_001166142.1 | 3 | 347 | ||
NM_025189.3 | NP_079465.3 | 3 | 347 | |||
GenBank mRNA | AK023721.1 | - | 3 | 327 | ||
AK290475.1 | - | 3 | 347 | |||
AY269787.1 | - | 3 | 276 | |||
BC111776.1 | - | 3 | 235 | 40033720 | ||
DB255551.1 | - | 3 | 310 |