Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038108.1 | 4 | 1167 | |||
NR_038109.1 | 4 | 1163 | ||||
NR_038110.1 | 3 | 1068 | ||||
NR_038111.1 | 3 | 1064 | ||||
GenBank mRNA | AB073602.1 | - | 4 | 1027 | ||
AK024090.1 | - | 2 | 507 | |||
AK055935.1 | - | 5 | 1095 | |||
AK129904.1 | - | 4 | 1033 | |||
BC042949.2 | - | 3 | 2236 | |||
BC112893.1 | - | 2 | 840 | |||
BX647993.1 | - | 3 | 1023 |
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127660.1 | - | 1 | 452 |
Gene Symbol: FBXO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018438.5 | NP_060908.1 | 6 | 1298 | ||
XM_005263448.4 | XP_005263505.1 | 6 | 1238 | |||
XM_005263449.4 | XP_005263506.1 | 6 | 1185 | |||
XM_005263451.4 | XP_005263508.1 | 6 | 1238 | |||
XM_006710571.3 | XP_006710634.1 | 6 | 1266 | |||
XM_011541232.2 | XP_011539534.1 | 6 | 1232 | |||
XM_017001007.1 | XP_016856496.1 | 7 | 1300 | |||
GenBank mRNA | BC020880.1 | - | 6 | 1240 | 4614957 |
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC006408.1 | - | 4 | 372 |