Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171816.1 | NP_001165287.1 | 2 | 944 | ||
NM_178841.3 | NP_849163.1 | 2 | 367 | |||
XM_011522845.2 | XP_011521147.1 | 2 | 361 | |||
XM_011522846.1 | XP_011521148.1 | 2 | 367 | |||
XM_017022910.1 | XP_016878399.1 | 1 | 2952 | |||
XM_017022911.1 | XP_016878400.1 | 2 | 361 | |||
XM_017022912.1 | XP_016878401.1 | 2 | 361 | |||
XM_017022913.1 | XP_016878402.1 | 2 | 1598 | |||
GenBank mRNA | AK057106.1 | - | 2 | 944 | ||
BC013948.2 | - | 2 | 293 | 3509621 | ||
BC017226.1 | - | 2 | 345 | 4299817 | ||
DA376321.1 | - | 2 | 367 |