Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171815.1 | NP_001165286.1 | 4 | 418 | ||
NM_001171816.1 | NP_001165287.1 | 5 | 1267 | |||
NM_178841.3 | NP_849163.1 | 5 | 690 | |||
XM_011522845.2 | XP_011521147.1 | 6 | 841 | |||
XM_011522846.1 | XP_011521148.1 | 5 | 690 | |||
XM_011522847.1 | XP_011521149.1 | 4 | 442 | |||
XM_017022910.1 | XP_016878399.1 | 4 | 4483 | |||
XM_017022913.1 | XP_016878402.1 | 5 | 1921 | |||
GenBank mRNA | AI800509.1 | - | 2 | 261 | ||
AK057106.1 | - | 5 | 1267 | |||
AK057201.1 | - | 4 | 338 | |||
AK290920.1 | - | 5 | 657 | |||
BC013948.2 | - | 5 | 616 | 3509621 | ||
BC017226.1 | - | 5 | 668 | 4299817 | ||
BI911983.1 | - | 4 | 425 |