Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152308.2 | NP_689521.1 | 2 | 1193 | ||
NR_130754.1 | 2 | 970 | ||||
GenBank mRNA | AK123764.1 | - | 2 | 1182 | ||
BC013040.2 | - | 2 | 1161 | 3542171 | ||
BC022427.1 | - | 2 | 1177 | 4252688 | ||
BC031016.1 | - | 2 | 1181 | 4659915 | ||
BC039361.1 | - | 5 | 1615 | 5270594 |