Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198273.2 | NP_938014.1 | 3 | 436 | ||
XM_005248421.4 | XP_005248478.1 | 3 | 901 | |||
GenBank mRNA | AK291478.1 | - | 3 | 433 | ||
AL832721.1 | - | 3 | 417 | |||
BC136740.1 | - | 3 | 328 | 9020730 | ||
BC136741.1 | - | 3 | 328 | 9020731 | ||
BP873090.1 | - | 3 | 437 | |||
BX500166.1 | - | 3 | 445 | |||
BX537972.1 | - | 3 | 406 | |||
DN994993.1 | - | 3 | 445 |