Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300807.1 | NP_001287736.1 | 2 | 552 | ||
NM_001300808.1 | NP_001287737.1 | 2 | 433 | |||
NM_138456.3 | NP_612465.3 | 3 | 620 | |||
GenBank mRNA | AK092453.1 | - | 2 | 430 | ||
AK096702.1 | - | 3 | 620 | |||
AK300917.1 | - | 2 | 549 | |||
BC012330.1 | - | 2 | 410 | 4638025 | ||
BP206576.1 | - | 2 | 433 | |||
HM134154.1 | - | 3 | 620 |