Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282309.1 | NP_001269238.1 | 4, 5 | 432 | ||
NM_001282310.1 | NP_001269239.1 | 3, 4 | 347 | |||
NM_054020.3 | NP_473361.1 | 3, 4 | 533 | |||
NM_172095.2 | NP_742093.1 | 3, 4 | 533 | |||
NR_110319.1 | 2, 3 | 320 | ||||
XM_017021904.1 | XP_016877393.1 | 3, 4 | 539 | |||
GenBank mRNA | AF411817.1 | - | 2, 3 | 318 | ||
AF411818.1 | - | 2, 3 | 318 | |||
AF411819.1 | - | 2, 3 | 318 | |||
AK122607.1 | - | 4, 5 | 431 | |||
BC028728.1 | - | 3, 4 | 538 | |||
BC064387.1 | - | 3, 4 | 347 | 5744415 | ||
BX648099.1 | - | 3, 4 | 569 |