Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145201.1 | NP_001138673.1 | 2, 3 | 163 | ||
NM_145202.4 | NP_660203.3 | 2, 3 | 163 | |||
GenBank mRNA | AF123768.1 | - | 1, 2 | 92 | ||
AF421885.1 | - | 2, 3 | 105 | |||
AY158074.1 | - | 1, 2 | 93 | |||
AY358908.1 | - | 2, 3 | 142 | |||
BC029447.2 | - | 1, 2 | 78 | |||
BC061643.1 | - | 2, 3 | 97 | 5748898 | ||
BC071872.1 | - | 2, 3 | 96 | 6284088 | ||
BC093853.1 | - | 2, 3 | 135 | 7939698 | ||
BC101743.1 | - | 2, 3 | 135 | 8069249 | ||
BC143590.1 | - | 2, 3 | 135 | 9052101 | ||
BC143591.1 | - | 2, 3 | 135 | 9052102 | ||
BC143592.1 | - | 2, 3 | 135 | 9052103 | ||
DQ884405.1 | - | 1, 2 | 85 | |||
GU727628.1 | - | 2, 3 | 105 |
Gene Symbol: ZNF511-PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU937518.1 | - | 5, 6 | 584 |