Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183058.2 | NP_898881.2 | 3, 4 | 451 | ||
GenBank mRNA | AF139543.1 | - | 3, 4 | 430 | ||
AI637906.1 | - | 2, 3 | 323 | |||
BC066294.1 | - | 3, 4 | 451 | 4374284 |