Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008781.2 | NP_001008781.2 | 1 | 649 | ||
XM_017017178.1 | XP_016872667.1 | 2 | 765 | |||
XM_017017179.1 | XP_016872668.1 | 3 | 987 | |||
XM_017017180.1 | XP_016872669.1 | 2 | 764 | |||
XM_017017181.1 | XP_016872670.1 | 2 | 764 | |||
XM_017017182.1 | XP_016872671.1 | 2 | 764 | |||
XM_017017183.1 | XP_016872672.1 | 2 | 764 | |||
XM_017017184.1 | XP_016872673.1 | 2 | 763 |