Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174891.3 | NP_777551.2 | 5 | 1115 | ||
GenBank mRNA | AF038188.1 | - | 5 | 679 | ||
AK057249.1 | - | 6 | 1191 | |||
BC003539.1 | - | 4 | 427 | |||
BC011537.1 | - | 4 | 945 | |||
BC025175.1 | - | 5 | 626 | |||
BC137495.1 | - | 5 | 1058 | 9021495 | ||
BC144671.1 | - | 5 | 1044 |