Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 1 | 414 | ||
NM_203467.1 | NP_982292.1 | 1 | 414 | |||
NR_037792.1 | 1 | 414 | ||||
NR_037793.1 | 1 | 414 | ||||
GenBank mRNA | AK293156.1 | - | 1 | 133 | ||
BC030142.2 | - | 1 | 350 | |||
BC064133.1 | - | 1 | 159 | |||
BC093697.1 | - | 1 | 116 | |||
BC107888.1 | - | 1 | 163 | |||
BC112241.1 | - | 1 | 116 | |||
BC139921.1 | - | 1 | 154 | 40148567 | ||
BI825814.1 | - | 1 | 341 | |||
BI829295.1 | - | 1 | 351 | |||
BQ059807.1 | - | 1 | 84 | |||
BX248298.1 | - | 1 | 157 | |||
BX648029.1 | - | 1 | 337 |
Gene Symbol: LOC100288910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX161511.1 | - | 1 | 51 |