Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083614.1 | NP_001077083.1 | 9 | 1534 | ||
NR_003501.1 | 9 | 1534 | ||||
XM_011545738.1 | XP_011544040.1 | 9 | 1824 | |||
XM_011545739.1 | XP_011544041.1 | 9 | 1443 | |||
GenBank mRNA | AB075850.1 | - | 9 | 1491 | ||
AK095998.1 | - | 9 | 1523 | |||
AL832489.1 | - | 9 | 1510 | |||
BX648129.1 | - | 7 | 1213 |