Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145255.3 | NP_660298.2 | 4, 5 | 563 | ||
NM_148887.2 | NP_683685.1 | 5, 6 | 661 | |||
NR_037575.1 | 4, 5 | 712 | ||||
GenBank mRNA | AK127167.1 | - | 4, 5 | 1438 | ||
AK293624.1 | - | 4, 5 | 701 | |||
AL558270.3 | - | 5, 6 | 626 | |||
BC015904.1 | - | 4, 5 | 534 | 3919892 | ||
BC039852.1 | - | 4, 5 | 542 | |||
BC052601.1 | - | 4, 5 | 552 | 6085551 |