Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145255.3 | NP_660298.2 | 5 | 790 | ||
NM_148887.2 | NP_683685.1 | 6 | 888 | |||
NR_037575.1 | 5 | 939 | ||||
GenBank mRNA | AK127167.1 | - | 5 | 1665 | ||
AK293624.1 | - | 5 | 928 | |||
BC015904.1 | - | 5 | 761 | 3919892 | ||
BC039852.1 | - | 5 | 769 | |||
BC052601.1 | - | 5 | 779 | 6085551 |