Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFMID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010982.4 | NP_001010982.2 | 3 | 237 | ||
NM_001145526.2 | NP_001138998.1 | 3 | 237 | |||
XM_011524327.2 | XP_011522629.1 | 4 | 405 | |||
XM_011524328.2 | XP_011522630.1 | 4 | 377 | |||
XM_011524329.2 | XP_011522631.1 | 4 | 419 | |||
XM_011524330.2 | XP_011522632.1 | 4 | 423 | |||
XM_011524331.2 | XP_011522633.1 | 4 | 405 | |||
XM_011524332.2 | XP_011522634.1 | 4 | 377 | |||
XM_011524333.2 | XP_011522635.1 | 3 | 196 | |||
XM_011524334.2 | XP_011522636.1 | 4 | 405 | |||
XM_011524335.2 | XP_011522637.1 | 4 | 405 | |||
XM_011524336.2 | XP_011522638.1 | 3 | 196 | |||
XM_011524337.2 | XP_011522639.1 | 3 | 238 | |||
GenBank mRNA | BC132824.1 | - | 3 | 237 | 40146846 | |
BC142973.1 | - | 3 | 192 | |||
BX648442.1 | - | 3 | 193 |