Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027159.1 | 2, 3 | 487 | |||
NR_027160.1 | 3, 4 | 610 | ||||
NR_027162.1 | 3, 4 | 663 | ||||
NR_027163.1 | 4, 5 | 397 | ||||
NR_027164.1 | 4, 5 | 344 | ||||
NR_027166.1 | 3, 4 | 221 | ||||
NR_027168.2 | 4, 5 | 313 | ||||
NR_027170.1 | 4, 5 | 265 | ||||
NR_027171.1 | 2, 3 | 487 | ||||
NR_027172.2 | 3, 4 | 190 | ||||
NR_027173.1 | 3, 4 | 221 | ||||
NR_027174.1 | 3, 4 | 221 | ||||
NR_027175.1 | 3, 4 | 221 | ||||
NR_027177.1 | 3, 4 | 221 | ||||
NR_027178.1 | 3, 4 | 221 | ||||
NR_027179.1 | 3, 4 | 221 | ||||
NR_027667.1 | 3, 4 | 531 | ||||
NR_045021.1 | 3, 4 | 200 | ||||
NR_045022.1 | 4, 5 | 339 | ||||
NR_045024.1 | 2, 3 | 487 | ||||
NR_045025.1 | 2, 3 | 133 | ||||
NR_045026.1 | 4, 5 | 397 | ||||
NR_045029.1 | 3, 4 | 295 | ||||
GenBank mRNA | AK098643.1 | - | 3, 4 | 171 | ||
AK307982.1 | - | 3, 4 | 613 | |||
AK311247.1 | - | 3, 4 | 777 | |||
BC019833.1 | - | 2, 3 | 147 | |||
BC027986.1 | - | 3, 4 | 481 | 4940349 | ||
BC040159.1 | - | 3, 4 | 460 | 5539041 | ||
BQ070126.1 | - | 2, 3 | 225 | |||
BU506593.1 | - | 328 | ||||
BU599615.1 | - | 4, 5 | 351 | |||
BX538291.1 | - | 4, 5 | 216 | |||
CB150672.1 | - | 4, 5 | 294 | |||
CD050014.1 | - | 3, 4 | 194 | |||
CD389053.1 | - | 3, 4 | 171 | |||
CF126952.1 | - | 2, 3 | 403 | |||
CR989654.1 | - | 3, 4 | 274 | |||
DA131201.1 | - | 3, 4 | 171 | |||
DA193585.1 | - | 3, 4 | 356 | |||
DA383448.1 | - | 3, 4 | 171 | |||
DA440445.1 | - | 4, 5 | 290 | |||
DA485648.1 | - | 3, 4 | 172 | |||
DA494627.1 | - | 2, 3 | 438 | |||
DA793334.1 | - | 2, 3 | 87 | |||
DA802938.1 | - | 3, 4 | 171 | |||
DA857697.1 | - | 3, 4 | 171 | |||
DA867958.1 | - | 4, 5 | 349 | |||
DB258221.1 | - | 2, 3 | 438 | |||
DW409012.1 | - | 3, 4 | 188 |