Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042461.2 | NP_001035926.1 | 2 | 466 | ||
NM_001042462.1 | NP_001035927.1 | 2 | 318 | |||
NM_174894.2 | NP_777554.1 | 2 | 489 | |||
GenBank mRNA | AK292001.1 | - | 2 | 296 | ||
BC042161.1 | - | 2 | 435 | 4869377 | ||
BG528980.1 | - | 2 | 319 | |||
BI196928.1 | - | 2 | 410 | |||
BM841051.1 | - | 2 | 306 | |||
BU736689.1 | - | 1 | 377 |