Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282349.1 | NP_001269278.1 | 5 | 770 | ||
NM_001282350.1 | NP_001269279.1 | 4 | 737 | |||
NM_130771.4 | NP_570127.3 | 6 | 782 | |||
NM_133168.4 | NP_573398.2 | 4 | 737 | |||
NM_133169.4 | NP_573399.2 | 5 | 770 | |||
NM_206818.2 | NP_996554.2 | 6 | 782 | |||
GenBank mRNA | AF251702.1 | - | 4 | 624 | ||
AF391162.1 | - | 4 | 585 | |||
AF391163.1 | - | 5 | 590 | |||
AF391164.1 | - | 3 | 583 | |||
AF474152.1 | - | 4 | 616 | |||
AF474153.1 | - | 3 | 545 | |||
AK130199.1 | - | 5 | 614 | |||
AK311307.1 | - | 4 | 710 | |||
BC035023.2 | - | 5 | 771 | 4825982 |