Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145807.2 | NP_665806.1 | 2 | 334 | ||
XM_011526443.2 | XP_011524745.1 | 2 | 372 | |||
GenBank mRNA | BC018654.2 | - | 2 | 260 | 2961115 | |
BC018697.2 | - | 2 | 260 | 2961115 | ||
BC021210.2 | - | 2 | 280 | 4137205 | ||
BC033207.1 | - | 2 | 260 | 4903484 |