Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243079.1 | NP_001230008.1 | 13, 14 | 1535 | ||
GenBank mRNA | AK054808.1 | - | 13, 14 | 1535 | ||
AK122648.1 | - | 12, 13 | 1707 | |||
BC100791.2 | - | 10, 11 | 1099 | |||
BC100792.1 | - | 10, 11 | 1099 |