Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080605.3 | NP_542172.2 | 1 | 813 | ||
GenBank mRNA | BC051719.1 | - | 1 | 709 | ||
BF002665.1 | - | 1 | 218 |