Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 1 | 610 | ||
NM_152366.4 | NP_689579.3 | 1 | 610 | |||
NR_033385.1 | 2 | 379 | ||||
NR_033386.1 | 2 | 379 | ||||
GenBank mRNA | AK093268.1 | - | 1 | 599 | ||
AY601914.1 | - | 1 | 609 | |||
BC022077.1 | - | 1 | 609 | 4822843 | ||
BC032482.1 | - | 1 | 628 | 5222141 | ||
BC066896.1 | - | 1 | 604 | 4828667 | ||
BG718418.1 | - | 1 | 608 | |||
BI549017.1 | - | 2 | 382 | |||
BP379202.1 | - | 2 | 380 | |||
BQ638407.1 | - | 1 | 348 | |||
BU173744.1 | - | 2 | 356 |