Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 3 | 864 | ||
NM_152366.4 | NP_689579.3 | 3 | 845 | |||
NR_033385.1 | 4 | 614 | ||||
NR_033386.1 | 4 | 633 | ||||
GenBank mRNA | AK093268.1 | - | 1 | 1209 | ||
AY601914.1 | - | 3 | 863 | |||
BC022077.1 | - | 3 | 844 | 4822843 | ||
BC032482.1 | - | 1 | 1238 | 5222141 | ||
BC066896.1 | - | 3 | 839 | 4828667 | ||
BI549017.1 | - | 3 | 457 | |||
BQ638407.1 | - | 3 | 583 | |||
BU173744.1 | - | 4 | 610 | |||
BU622692.1 | - | 2 | 488 |