Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195736.1 | NP_001182665.1 | 6, 7 | 771 | ||
NM_001195737.1 | NP_001182666.1 | 6, 7 | 705 | |||
NM_001195738.1 | NP_001182667.1 | 6, 7 | 685 | |||
NM_001195740.1 | NP_001182669.1 | 5, 6 | 609 | |||
NM_001195741.1 | NP_001182670.1 | 5, 6 | 606 | |||
NM_152371.3 | NP_689584.2 | 6, 7 | 717 | |||
NR_036637.1 | 6, 7 | 650 | ||||
NR_036638.1 | 6, 7 | 618 | ||||
XM_006710354.3 | XP_006710417.1 | 6, 7 | 705 | |||
XM_011540664.1 | XP_011538966.1 | 6, 7 | 761 | |||
XM_011540665.1 | XP_011538967.1 | 6, 7 | 729 | |||
XM_011540666.1 | XP_011538968.1 | 6, 7 | 709 | |||
GenBank mRNA | AF425266.1 | - | 6, 7 | 661 | ||
AK057027.1 | - | 6, 7 | 702 | |||
AK075273.1 | - | 6, 7 | 637 | |||
AK094901.1 | - | 5, 6 | 519 | |||
AK291908.1 | - | 6, 7 | 643 | |||
AK298926.1 | - | 5, 6 | 560 | |||
AK303504.1 | - | 6, 7 | 677 | |||
AK308034.1 | - | 6, 7 | 627 | |||
AK316243.1 | - | 5, 6 | 519 | |||
BC022547.1 | - | 6, 7 | 657 | 4812314 | ||
BI908668.1 | - | 6, 7 | 537 | |||
BQ709693.1 | - | 553 | ||||
CR749706.1 | - | 1, 2 | 449 |