Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080825.3 | NP_543015.1 | 1 | 166 | ||
GenBank mRNA | BC032818.1 | - | 1 | 116 | 5172528 | |
BX378327.2 | - | 1 | 166 | |||
DQ080434.1 | - | 1 | 168 |
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124388.1 | - | 1 | 324 |