Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138797.2 | NP_620152.1 | 1 | 209 | ||
NR_036556.1 | 1 | 209 | ||||
XM_006724137.1 | XP_006724200.1 | 1 | 78 | |||
XM_011529877.2 | XP_011528179.1 | 1 | 500 | |||
XM_011529883.2 | XP_011528185.1 | 1 | 232 | |||
XM_017028593.1 | XP_016884082.1 | 1 | 479 | |||
GenBank mRNA | BC014641.1 | - | 1 | 159 | 2988073 | |
BC066909.1 | - | 1 | 176 | 4795922 | ||
BX401122.2 | - | 1 | 208 | |||
CD300696.1 | - | 108 | ||||
CR456471.1 | - | 1 | 169 |