Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080824.2 | NP_001074293.1 | 2 | 652 | ||
NM_001277053.1 | NP_001263982.1 | 2 | 652 | |||
NM_001307978.1 | NP_001294907.1 | 2 | 652 | |||
XM_011532504.1 | XP_011530806.1 | 2 | 452 | |||
GenBank mRNA | AK296608.1 | - | 2 | 489 | ||
AK309615.1 | - | 2 | 491 | |||
BC051789.1 | - | 2 | 653 | |||
JQ653415.1 | - | 2 | 446 |