Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029881.2 | NP_001025052.1 | 5, 6 | 496 | ||
XM_011532514.2 | XP_011530816.1 | 5, 6 | 579 | |||
XM_011532517.2 | XP_011530819.1 | 3, 4 | 866 | |||
XM_017003329.1 | XP_016858818.1 | 4, 5 | 1350 | |||
XM_017003330.1 | XP_016858819.1 | 3, 4 | 1260 | |||
GenBank mRNA | AW138422.1 | - | 2, 3 | 291 | ||
BC127684.1 | - | 5, 6 | 496 | 40127392 | ||
BC127685.1 | - | 5, 6 | 494 | 40127393 | ||
BC140923.1 | - | 5, 6 | 470 | 9056735 |